Assay Details
| Gene Symbol: SLC46A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024519.1 | NP_001019690.1 | 1 - 2 | 1231 | 76 | |
| GenBank mRNA | BT021767.1 | - | 1 - 2 | 1231 | 76 | |
More Information
Set Membership: |
|

SLC46A2
Genomic Map