Assay Details
| Gene Symbol: DCAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024520.1 | NP_001019691.1 | 13 - 14 | 1365 | 61 | |
| GenBank mRNA | BC116026.1 | - | 13 - 14 | 1385 | 61 | |
| BT021675.1 | - | 13 - 14 | 1365 | 61 | ||
More Information
Set Membership: |
|

DCAF4
Genomic Map