Assay Details
| Gene Symbol: GPAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024529.1 | NP_001019700.1 | 11 - 12 | 1626 | 84 | |
| GenBank mRNA | BC102317.1 | - | 11 - 12 | 1631 | 84 | |
| BT021804.1 | - | 11 - 12 | 1626 | 84 | ||
More Information
Set Membership: |
|

GPAA1
Genomic Map