Assay Details
| Gene Symbol: REG3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024534.3 | NP_001019705.2 | 1 - 2 | 54 | 113 | |
| GenBank mRNA | BC102114.1 | - | 1 - 2 | 57 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102114.1, NM_001024534.3 transcripts.
Set Membership: |
|

REG3G
Genomic Map