Assay Details
| Gene Symbol: HDHD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024538.1 | NP_001019709.1 | 3 - 4 | 716 | 68 | |
| GenBank mRNA | BC103273.1 | - | 4 - 5 | 540 | 68 | |
| BT021739.1 | - | 3 - 4 | 716 | 68 | ||
More Information
Set Membership: |
|

HDHD5
Genomic Map