Assay Details
| Gene Symbol: BCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024559.1 | NP_001019730.1 | 6 - 7 | 975 | 59 | |
| GenBank mRNA | DQ008469.1 | - | 6 - 7 | 975 | 59 | |
More Information
Set Membership: |
|

BCO1
Genomic Map