Assay Details
| Gene Symbol: DSTYK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024824.1 | NP_001019995.1 | 2 - 3 | 721 | 59 | |
| GenBank mRNA | DQ054504.1 | - | 2 - 3 | 721 | 59 | |
More Information
Set Membership: |
|

DSTYK
Genomic Map