Assay Details
| Gene Symbol: BHLHE40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024929.1 | NP_001020100.1 | 3 - 4 | 485 | 70 | |
| GenBank mRNA | BT020754.1 | - | 3 - 4 | 485 | 70 | |
More Information
Set Membership: |
|

BHLHE40
Genomic Map