Assay Details
| Gene Symbol: TWF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025320.2 | NP_001020491.1 | 3 - 4 | 324 | 65 | |
| GenBank mRNA | AY911376.1 | - | 3 - 4 | 325 | 65 | |
More Information
Set Membership: |
|

TWF1
Genomic Map