Assay Details
| Gene Symbol: EIF1AX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025329.2 | NP_001020500.1 | 3 - 4 | 369 | 79 | |
| GenBank mRNA | AY911362.1 | - | 3 - 4 | 370 | 79 | |
| BC109650.1 | - | 3 - 4 | 428 | 79 | ||
More Information
Set Membership: |
|

EIF1AX
Genomic Map