Assay Details
| Gene Symbol: RBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025343.1 | NP_001020514.1 | 1 - 2 | 198 | 77 | |
| GenBank mRNA | AF502256.1 | - | 1 - 2 | 198 | 77 | |
| BC109589.1 | - | 1 - 2 | 117 | 77 | ||
| BT021637.1 | - | 5 - 6 | 888 | 77 | ||
More Information
Set Membership: |
|

RBP1
Genomic Map