Assay Details
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025346.1 | NP_001020517.1 | 1 - 2 | 105 | 90 | |
GenBank mRNA | BC133652.1 | - | 1 - 2 | 117 | 90 | |
DQ062556.1 | - | 1 - 2 | 105 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
