Assay Details
| Gene Symbol: SPINK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025348.1 | NP_001020519.1 | 1 - 2 | 110 | 64 | |
| GenBank mRNA | AY911325.1 | - | 1 - 2 | 110 | 64 | |
| BC102485.1 | - | 1 - 2 | 110 | 64 | ||
More Information
Set Membership: |
|

SPINK1
Genomic Map