Assay Details
| Gene Symbol: CLCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031757.1 | NP_001026927.1 | 11 - 12 | 2266 | 58 | |
| GenBank mRNA | BC151324.1 | - | 10 - 11 | 2045 | 58 | |
| DQ102705.1 | - | 11 - 12 | 2266 | 58 | ||
More Information
Set Membership: |
|

CLCN4
Genomic Map