Assay Details
| Gene Symbol: RHEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031764.2 | NP_001026934.1 | 6 - 7 | 432 | 94 | |
| GenBank mRNA | AY911379.1 | - | 6 - 7 | 433 | 94 | |
| BC102507.1 | - | 1 | 476 | 94 | ||
More Information
Set Membership: |
|

RHEB
Genomic Map