Assay Details
| Gene Symbol: RAB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033628.1 | NP_001028800.1 | 3 - 4 | 376 | 74 | |
| GenBank mRNA | BC103436.1 | - | 3 - 4 | 376 | 74 | |
| BT029815.1 | - | 4 - 5 | 464 | 74 | ||
More Information
Set Membership: |
|

RAB1A
Genomic Map