Assay Details
| Gene Symbol: FHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034223.2 | NP_001029395.1 | 3 - 4 | 516 | 84 | |
| GenBank mRNA | BC103276.1 | - | 3 - 4 | 517 | 84 | |
More Information
Set Membership: |
|

FHL3
Genomic Map