Assay Details
| Gene Symbol: SPC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034230.2 | NP_001029402.1 | 6 - 7 | 680 | 64 | |
| GenBank mRNA | BC103246.1 | - | 6 - 7 | 682 | 64 | |
More Information
Set Membership: |
|

SPC25
Genomic Map