Assay Details
| Gene Symbol: FBXL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034252.1 | NP_001029424.1 | 4 - 5 | 772 | 127 | |
| GenBank mRNA | BC103468.1 | - | 4 - 5 | 772 | 127 | |
More Information
Set Membership: |
|

FBXL21
Genomic Map