Assay Details
| Gene Symbol: SCNM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034254.2 | NP_001029426.1 | 4 - 5 | 373 | 88 | |
| GenBank mRNA | BC103163.1 | - | 4 - 5 | 374 | 88 | |
More Information
Set Membership: |
|

SCNM1
Genomic Map