Assay Details
| Gene Symbol: C7H1orf35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034272.1 | NP_001029444.1 | 7 - 8 | 708 | 65 | |
| GenBank mRNA | BT021507.1 | - | 7 - 8 | 708 | 65 | |
More Information
Set Membership: |
|

C7H1orf35
Genomic Map