Assay Details
| Gene Symbol: ICOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034275.1 | NP_001029447.1 | 4 - 5 | 591 | 66 | |
| GenBank mRNA | BC149958.1 | - | 3 - 4 | 1190 | 66 | |
| BT021638.1 | - | 4 - 5 | 591 | 66 | ||
More Information
Set Membership: |
|

ICOS
Genomic Map