Assay Details
| Gene Symbol: DEGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034289.2 | NP_001029461.1 | 1 - 2 | 179 | 141 | |
| GenBank mRNA | BC103031.1 | - | 1 - 2 | 179 | 141 | |
More Information
Set Membership: |
|

DEGS1
Genomic Map