Assay Details
| Gene Symbol: HAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034347.1 | NP_001029519.1 | 6 - 7 | 672 | 86 | |
| GenBank mRNA | BC120387.1 | - | 6 - 7 | 643 | 86 | |
| BT021536.1 | - | 6 - 7 | 672 | 86 | ||
More Information
Set Membership: |
|

HAT1
Genomic Map