Assay Details
| Gene Symbol: TMEM54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034364.2 | NP_001029536.1 | 1 - 2 | 163 | 77 | |
| GenBank mRNA | BC102511.1 | - | 1 - 2 | 164 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102511.1, NM_001034364.2 transcripts.
Set Membership: |
|

TMEM54
Genomic Map