Assay Details
| Gene Symbol: CTSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034435.1 | NP_001029607.1 | 4 - 5 | 528 | 65 | |
| GenBank mRNA | BC109853.1 | - | 4 - 5 | 455 | 65 | |
| BT021052.1 | - | 4 - 5 | 528 | 65 | ||
More Information
Set Membership: |
|

CTSK
Genomic Map