Assay Details
Gene Symbol: TRABD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034452.1 | NP_001029624.1 | 4 - 5 | 469 | 82 | |
GenBank mRNA | BC149347.1 | - | 5 - 6 | 507 | 82 | |
BT021644.1 | - | 4 - 5 | 469 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
