Assay Details
| Gene Symbol: CFAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034453.2 | NP_001029625.1 | 9 - 10 | 947 | 73 | |
| GenBank mRNA | BC102896.1 | - | 9 - 10 | 948 | 73 | |
More Information
Set Membership: |
|

CFAP36
Genomic Map