Assay Details
| Gene Symbol: SLC25A37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034549.1 | NP_001029721.1 | 1 - 2 | 349 | 65 | |
| GenBank mRNA | BC103255.1 | - | 1 - 2 | 349 | 65 | |
More Information
Set Membership: |
|

SLC25A37
Genomic Map