Assay Details
| Gene Symbol: PELO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034566.1 | NP_001029738.1 | 1 - 2 | 988 | 80 | |
| GenBank mRNA | BC118458.1 | - | 1 - 2 | 1030 | 80 | |
| BT021497.1 | - | 1 - 2 | 988 | 80 | ||
| BT025367.1 | - | 1 - 2 | 1004 | 80 | ||
More Information
Set Membership: |
|

PELO
Genomic Map