Assay Details
| Gene Symbol: TAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034590.1 | NP_001029762.1 | 7 - 8 | 798 | 84 | |
| GenBank mRNA | BC146179.1 | - | 7 - 8 | 814 | 84 | |
| BT021798.1 | - | 7 - 8 | 798 | 84 | ||
More Information
Set Membership: |
|

TAT
Genomic Map