Assay Details
| Gene Symbol: PRXL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034599.2 | NP_001029771.1 | 5 - 6 | 776 | 126 | |
| GenBank mRNA | BC103249.1 | - | 5 - 6 | 777 | 126 | |
More Information
Set Membership: |
|

PRXL2A
Genomic Map