Assay Details
| Gene Symbol: CARD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034753.1 | NP_001029925.1 | 1 - 2 | 235 | 69 | |
| GenBank mRNA | BC112579.1 | - | 1 - 2 | 99 | 69 | |
| BT021706.1 | - | 1 - 2 | 235 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112579.1, NM_001034753.1 transcripts.
Set Membership: |
|

CARD19
Genomic Map