Assay Details
| Gene Symbol: MED10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034756.2 | NP_001029928.1 | 2 - 3 | 265 | 86 | |
| GenBank mRNA | BC102439.1 | - | 2 - 3 | 266 | 86 | |
More Information
Set Membership: |
|

MED10
Genomic Map