Assay Details
| Gene Symbol: SH3BGRL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034763.2 | NP_001029935.1 | 1 - 2 | 148 | 73 | |
| GenBank mRNA | BC102048.1 | - | 1 - 2 | 149 | 73 | |
More Information
Set Membership: |
|

SH3BGRL3
Genomic Map