Assay Details
| Gene Symbol: POP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034770.1 | NP_001029942.1 | 4 - 5 | 850 | 59 | |
| NM_001113282.2 | NP_001106753.1 | 4 - 5 | 595 | 59 | ||
| GenBank mRNA | BC112695.1 | - | 4 - 5 | 596 | 59 | |
| BT021479.1 | - | 4 - 5 | 850 | 59 | ||
More Information
Set Membership: |
|

POP4
Genomic Map