Assay Details
| Gene Symbol: MRPL57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034787.2 | NP_001029959.1 | 1 - 2 | 55 | 72 | |
| GenBank mRNA | BC102996.1 | - | 1 - 2 | 56 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102996.1, NM_001034787.2 transcripts.
Set Membership: |
|

MRPL57
Genomic Map