Assay Details
| Gene Symbol: NMUR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035048.1 | NP_001030220.1 | 3 - 4 | 1032 | 94 | |
| GenBank mRNA | BT021833.1 | - | 3 - 4 | 1032 | 94 | |
More Information
Set Membership: |
|

NMUR2
Genomic Map