Assay Details
| Gene Symbol: SNRPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035065.2 | NP_001030237.1 | 2 - 3 | 208 | 152 | |
| GenBank mRNA | BC102987.1 | - | 2 - 3 | 209 | 152 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102987.1, NM_001035065.2 transcripts.
Set Membership: |
|

SNRPD1
Genomic Map