Assay Details
| Gene Symbol: EIF3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035296.2 | NP_001030373.1 | 11 - 12 | 1633 | 61 | |
| GenBank mRNA | BC102112.1 | - | 11 - 12 | 1634 | 61 | |
More Information
Set Membership: |
|

EIF3L
Genomic Map