Assay Details
| Gene Symbol: CDK2AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035320.1 | NP_001030397.1 | 2 - 3 | 232 | 53 | |
| GenBank mRNA | BC103221.1 | - | 2 - 3 | 262 | 53 | |
| BT021910.1 | - | 2 - 3 | 232 | 53 | ||
More Information
Set Membership: |
|

CDK2AP2
Genomic Map