Assay Details
| Gene Symbol: MRPS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035332.1 | NP_001030409.1 | 8 - 9 | 831 | 78 | |
| GenBank mRNA | BC109583.1 | - | 8 - 9 | 1184 | 78 | |
| BT021542.1 | - | 8 - 9 | 831 | 78 | ||
| BT026199.1 | - | 8 - 9 | 830 | 78 | ||
More Information
Set Membership: |
|

MRPS9
Genomic Map