Assay Details
| Gene Symbol: TNFRSF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035336.2 | NP_001030413.1 | 7 - 8 | 822 | 75 | |
| GenBank mRNA | BC102869.1 | - | 7 - 8 | 823 | 75 | |
More Information
Set Membership: |
|

TNFRSF9
Genomic Map