Assay Details
| Gene Symbol: SCAMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035426.1 | NP_001030503.1 | 5 - 6 | 625 | 89 | |
| GenBank mRNA | BC102055.1 | - | 5 - 6 | 625 | 89 | |
| BT021532.1 | - | 5 - 6 | 638 | 89 | ||
More Information
Set Membership: |
|

SCAMP3
Genomic Map