Assay Details
| Gene Symbol: PDLIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035455.1 | NP_001030532.1 | 6 - 7 | 907 | 60 | |
| GenBank mRNA | BC146213.1 | - | 6 - 7 | 928 | 60 | |
| BT020979.1 | - | 6 - 7 | 907 | 60 | ||
More Information
Set Membership: |
|

PDLIM1
Genomic Map