Assay Details
| Gene Symbol: FKBP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035456.2 | NP_001030533.1 | 4 - 5 | 500 | 101 | |
| GenBank mRNA | BC102338.1 | - | 4 - 5 | 448 | 101 | |
| BT021075.1 | - | 4 - 5 | 477 | 101 | ||
| EE356851.1 | - | 4 - 5 | 500 | 101 | ||
More Information
Set Membership: |
|

FKBP1A
Genomic Map