Assay Details
| Gene Symbol: CTNNBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037142.1 | NP_001032219.1 | 3 - 4 | 326 | 55 | |
| GenBank mRNA | BC151327.1 | - | 4 - 5 | 520 | 55 | |
| BT020888.1 | - | 3 - 4 | 326 | 55 | ||
More Information
Set Membership: |
|

CTNNBIP1
Genomic Map