Assay Details
| Gene Symbol: UTS2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040484.1 | NP_001035574.1 | 1 | 1252 | 142 | |
| GenBank mRNA | BT021614.1 | - | 1 | 1252 | 142 | |
More Information
Set Membership: |
|

UTS2R
Genomic Map