Assay Details
| Gene Symbol: METAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040493.1 | NP_001035583.1 | 9 - 10 | 1128 | 84 | |
| GenBank mRNA | AB098975.1 | - | 4 - 5 | 474 | 84 | |
| BC102824.1 | - | 9 - 10 | 1128 | 84 | ||
More Information
Set Membership: |
|

METAP2
Genomic Map