Assay Details
| Gene Symbol: EGR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040497.1 | NP_001035587.1 | 1 - 2 | 265 | 92 | |
| GenBank mRNA | AJ293900.2 | - | 1 - 2 | 321 | 92 | |
More Information
Set Membership: |
|

EGR4
Genomic Map